2,4,5-Trichlorothiophenol ≥97.0%

Supplier: TCI America

Synonyms: 1-Mercapto-2,4,5-trichlorobenzene, 2,4,5-Trichlorothiophenol, 2,4,5-Trichlorobenzenethiol, 2,4,5-Trichlorophenyl Mercaptan

T0728-10G T0728-25G
TCT0728-10GEA 295.93 CAD
TCT0728-10G TCT0728-25G
2,4,5-Trichlorothiophenol ≥97.0%
2,4,5-Trichlorothiophenol

CAS Number: 3773-14-6
MDL Number: MFCD00014447
Molecular Formula: C6H3Cl3S
Molecular Weight: 213.50
Purity/Analysis Method: >97.0% (GC,T)
Form: Crystal
Color: White
Boiling point (°C): 154
Melting point (°C): 114


Other Product Names / Synonyms: 2,4,5-Trichlorothiophenol, 2,4,5-Trichlorophenyl Mercaptan

Formula: C₆H₃Cl₃S
MW: 213.51
Boiling Pt: 154 °C
Melting Pt: 114 °C
MDL Number: MFCD00014447
CAS Number: 3773-14-6
UN: 3335
ADR: 9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3f5b4c00d79e1790576b4ba343a7a84364823760&vid=77aa9f7a3d7cf38d19cb12eff3839c29ac6dd429&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR