Trioctylphosphine oxide ≥95.0%
Supplier: TCI America
Synonyms:
TOPO, Tri-n-octylphosphine Oxide, Trioctylphosphinoxide
T0504-100G
T0504-25G
TCT0504-100GEA
176.96
CAD
TCT0504-100G
TCT0504-25G
Trioctylphosphine oxide ≥95.0%
Trioctylphosphine oxide
CAS Number: 78-50-2
MDL Number: MFCD00002083
Molecular Formula: C24H51OP
Molecular Weight: 386.64
Purity/Analysis Method: >95.0% (GC)
Form: Crystal
Color: White
Boiling point (°C): 238
Melting point (°C): 54
Flash Point (°C): 110
Other Product Names / Synonyms: TOPO
Formula:
C₂₄H₅₁OP Boiling Pt: 238 °C Melting Pt: 54 °C Flash Pt: 110 °C |
MDL Number:
MFCD00002083 CAS Number: 78-50-2 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...