Tetraphenylmethane ≥96.0% (by GC)

Supplier: TCI America

T0169-1G T0169-5G
TCT0169-1GEA 135.88 CAD
TCT0169-1G TCT0169-5G
Tetraphenylmethane ≥96.0% (by GC)
Tetraphenylmethane

CAS Number: 630-76-2
MDL Number: MFCD00014428
Molecular Formula: C25H20
Molecular Weight: 320.44
Purity/Analysis Method: >96.0% (GC)
Form: Crystal
Color: White
Boiling point (°C): 431
Melting point (°C): 283
Storage Temperature: 0-10°C

Formula: C₂₅H₂₀
Boiling Pt: 431 °C
Melting Pt: 283 °C
MDL Number: MFCD00014428
CAS Number: 630-76-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6695c9b0be083749909a4cb14b3a204e85b81201&vid=6695c9b0be083749909a4cb14b3a204e85b81201&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR