(R)-(+)-SEGPHOS® ≥99.0% (by HPLC)

Supplier: TCI America

S0930-1G S0930-200MG
TCS0930-1GEA 287.56 CAD
TCS0930-1G TCS0930-200MG
(R)-(+)-SEGPHOS® ≥99.0% (by HPLC)
(R)-5,5'-Bis(diphenylphosphino)-4,4'-bibenzo[d][1,3]dioxole

CAS Number: 244261-66-3
MDL Number: MFCD09753005
Molecular Formula: C38H28O4P2
Molecular Weight: 610.59
Purity/Analysis Method: >99.0% (HPLC)
Form: Crystal
Color: White
Melting point (°C): 172
Specific rotation [a]20/D: 11 deg (C=0.5, CHCl3)


Product Categories: Synthetic Organic Chemistry, Asymmetric Synthesis, Ligands, Ligands ([2+2+2]Cycloaddition Reaction)
Other Product Names / Synonyms: (R)-(+)-5,5'-Bis(diphenylphosphino)-4,4'-bi-1,3-benzodioxole, (R)-(+)-4,4'-Bis(diphenylphosphino)-3,3'-bi(1,2-methylenedioxybenzene)

Formula: C₃₈H₂₈O₄P₂
Melting Pt: 172 °C
MDL Number: MFCD09753005
CAS Number: 244261-66-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9245aa46003cb573cc6bd40c08289da7255f5018&vid=9245aa46003cb573cc6bd40c08289da7255f5018&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (HPLC)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR