(S)-(-)-SEGPHOS® ≥99.0% (by HPLC)

Supplier: TCI America

S0929-1G S0929-200MG
TCS0929-1GEA 267.02 CAD
TCS0929-1G TCS0929-200MG
(S)-(-)-SEGPHOS® ≥99.0% (by HPLC)
(S)-5,5'-Bis(diphenylphosphino)-4,4'-bi-1,3-benzodioxole

CAS Number: 210169-54-3
MDL Number: MFCD09753005
Molecular Formula: C38H28O4P2
Molecular Weight: 610.59
Purity/Analysis Method: >99.0% (HPLC)
Form: Crystal
Color: White
Melting point (°C): 217
Specific rotation [a]20/D: -11 deg (C=0.5, CHCl3)


Product Categories: Synthetic Organic Chemistry, Asymmetric Synthesis, Ligands, Ligands ([2+2+2]Cycloaddition Reaction)
Other Product Names / Synonyms: (S)-(-)-5,5'-Bis(diphenylphosphino)-4,4'-bi-1,3-benzodioxole, (S)-(-)-4,4'-Bis(diphenylphosphino)-3,3'-bi(1,2-methylenedioxybenzene)

Formula: C₃₈H₂₈O₄P₂
Melting Pt: 217 °C
MDL Number: MFCD09753005
CAS Number: 210169-54-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=812c8ce7e2f785db643ee397eb7150e3f2f3e4cf&vid=812c8ce7e2f785db643ee397eb7150e3f2f3e4cf&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR