Sulfadoxin ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
Synonyms:
4-Amino-N-(5,6-dimethoxypyrimidin-4-yl)benzenesulfonamide, sulphadoxine, 4-Amino-N-(5,6-dimethoxy-4-pyrimidinyl)benzenesulfonamide, sulfadoxine, 4-Amino-N-(5,6-dimethoxy-4-pyrimidinyl)benzenesulphonamide, 4-Amino-N-(5,6-dimethoxypyrimidin-4-yl)benzenesulphonamide
CAS Number: 2447-57-6
MDL Number: MFCD00792890
Molecular Formula: C12H14N4O4S
Molecular Weight: 310.33
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Color: White
Melting point (°C): 198
Product Categories: Bioscience, Synthetic Antimicrobial Ingredients, Antiprotozoal Ingredients
Other Product Names / Synonyms: 4-Amino-N-(5,6-dimethoxy-4-pyrimidinyl)benzenesulfonamide
Formula:
C₁₂H₁₄N₄O₄S MW: 310.33 g/mol Melting Pt: 198 °C |
MDL Number:
MFCD00792890 CAS Number: 2447-57-6 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...