Sodium-2-propylvalerate ≥98.0% (by titrimetric analysis)

Supplier: TCI America

S0894-100G S0894-25G
TCS0894-100GEA 268.6 CAD
TCS0894-100G TCS0894-25G
Sodium-2-propylvalerate ≥98.0% (by titrimetric analysis)
Sodium-2-propylvalerate

CAS Number: 1069-66-5
MDL Number: MFCD00078604
Molecular Formula: C8H16O2
Molecular Weight: 166.20
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Color: White


Product Categories: Bioscience, Inhibitors (for Pharmacological Research), Neurotransmitter Agent Ingredients, Voltage-dependent Calcium Channel (VDCC) Antagonists, Epigenetics Research Reagents
Other Product Names / Synonyms: Sodium 2-Propylpentanoate, Valproic Acid Sodium Salt, 2-Propylvaleric Acid Sodium Salt, Sodium Valproate, 2-Propylpentanoic Acid Sodium Salt

Formula: C₈H₁₅O₂Na
MDL Number: MFCD00078604
CAS Number: 1069-66-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0c39a6e2138be203c76120d476025a686de2ccba&vid=0c39a6e2138be203c76120d476025a686de2ccba&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR