Sulfo-SMCC (3-Sulfo-N-succinimidyl-4-(N-maleimidomethyl)cyclohexane-1-carboxylate sodium salt)
Supplier: TCI America
Synonyms:
Sodium 1-((4-((2,5-dioxo-2,5-dihydro-1H-pyrrol-1-yl)methyl)cyclohexane-1-carbonyl)oxy)-2,5-dioxopyrrolidine-3-sulphonate, Sulphosuccinimidyl-trans-4-(N-maleimidomethyl)cyclohexane-1-carboxylate, Sodium 1-((4-((2,5-dioxo-2,5-dihydro-1H-pyrrol-1-yl)methyl)cyclohexane-1-carbonyl)oxy)-2,5-dioxopyrrolidine-3-sulfonate, Sulpho-SMCC Sodium Salt, SMCC, Water-Soluble, 4-(N-Maleimidomethyl)cyclohexane-1-carboxylic acid 3-sulpho-N-hydroxysuccinimide ester sodium salt, sulfo-SMCC Sodium Salt, sulpho-SMCC, 4-(N-Maleimidomethyl)cyclohexane-1-carboxylic acid 3-sulfo-N-hydroxysuccinimide ester sodium salt, Sulfosuccinimidyl-trans-4-(N-maleimidomethyl)cyclohexane-1-carboxylate, Sulfo-SMCC
CAS Number: 92921-24-9
MDL Number: MFCD00054978
Molecular Formula: C16H18N2O9S
Molecular Weight: 436.37
Form: Crystal
Color: White
Storage Temperature: <0°C
Other Product Names / Synonyms: Sulfo-SMCC Sodium Salt
Formula:
C₁₆H₁₈N₂O₉S MW: 436.37 g/mol |
MDL Number:
MFCD00054978 CAS Number: 92921-24-9 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...