Formaldehyde sodium bisulfite hemihydrate ≥97.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: Sodium Hydroxymethanesulphonate, Sodium Hydroxymethanesulfonate, Hydroxymethanesulfonic acid sodium salt, Hydroxymethanesulphonic acid sodium salt

S0880-25G S0880-500G
TCS0880-25GEA 26.86 CAD
TCS0880-25G TCS0880-500G
Formaldehyde sodium bisulfite hemihydrate ≥97.0% (by titrimetric analysis)
Formaldehyde sodium bisulfite hemihydrate

CAS Number: 63148-74-3
Molecular Formula: CH4O4S
Molecular Weight: 134.08
Purity/Analysis Method: >97.0% (T)
Form: Crystal
Color: White


Other Product Names / Synonyms: Hydroxymethanesulfonic Acid Sodium Salt, Sodium Hydroxymethanesulfonate

Formula: CH₄O₄S
MW: 134.08 g/mol
CAS Number: 63148-74-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cd0c8c3dac022e319e7e35f10908a197772a7255&vid=cd0c8c3dac022e319e7e35f10908a197772a7255&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR