Sodium stearyl fumarate ≥98.0%
Supplier: TCI America
Synonyms:
Fumaric Acid Stearyl Ester Sodium Salt, Fumaric Acid Octadecyl Ester Sodium Salt, Sodium Octadecyl Fumarate
S0879-100G
S0879-25G
TCS0879-100GEA
347.13
CAD
TCS0879-100G
TCS0879-25G
Sodium stearyl fumarate ≥98.0%
Sodium stearyl fumarate
CAS Number: 4070-80-8
MDL Number: MFCD00236640
Molecular Formula: C22H40O4
Molecular Weight: 390.54
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Color: White
Other Product Names / Synonyms: Fumaric Acid Stearyl Ester Sodium Salt, Sodium Octadecyl Fumarate, Fumaric Acid Octadecyl Ester Sodium Salt
Formula:
C₂₂H₄₀O₄ |
MDL Number:
MFCD00236640 CAS Number: 4070-80-8 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...