N-Succinimidyl Acetate 98,0

Supplier: TCI America

Synonyms: N-Acetoxysuccinimide, Ac-Osu, Acetic Acid N-Succinimidyl Ester, 2,5-Dioxopyrrolidin-1-yl acetate

S0878-25G S0878-5G
TCS0878-25GEA 327.22 CAD
TCS0878-25G TCS0878-5G
N-Succinimidyl Acetate 98,0
N-Succinimidyl Acetate

CAS Number: 14464-29-0
MDL Number: MFCD00037916
Molecular Formula: C6H7NO4
Molecular Weight: 157.13
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Color: White
Melting point (°C): 134
Storage Temperature: 0-10°C


Other Product Names / Synonyms: Acetic Acid N-Succinimidyl Ester, N-Acetoxysuccinimide

Formula: C₆H₇NO₄
MW: 157.13 g/mol
Melting Pt: 134 °C
MDL Number: MFCD00037916
CAS Number: 14464-29-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=517494412923cab442fa71246086ab5ae9551e68&vid=517494412923cab442fa71246086ab5ae9551e68&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR