Sodium ibandronate ≥90.0% (by titrimetric analysis)

Supplier: TCI America

S0877-1G S0877-5G
TCS0877-1GEA 353.24 CAD
TCS0877-1G TCS0877-5G
Sodium ibandronate ≥90.0% (by titrimetric analysis)
Sodium ibandronate

CAS Number: 138844-81-2
MDL Number: MFCD07197214
Molecular Formula: C9H23NO7P2
Molecular Weight: 341.21
Purity/Analysis Method: >90.0% (T)
Form: Crystal
Color: White
Storage Temperature: 0-10°C


Product Categories: Bioscience, Anti-osteoporosis Ingredients
Other Product Names / Synonyms: Ibandronic Acid Sodium Salt

Formula: C₉H₂₃NO₇P₂
MDL Number: MFCD07197214
CAS Number: 138844-81-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fcac9a0d4edab48e6c42bdf390cb2f49ae2a17f1&vid=fcac9a0d4edab48e6c42bdf390cb2f49ae2a17f1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 90.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR