Suplatast tosylate ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America

S0875-100MG S0875-1G
TCS0875-100MGEA 74.26 CAD
TCS0875-100MG TCS0875-1G
Suplatast tosylate ≥98.0% (by HPLC, total nitrogen)
Suplatast tosylate

CAS Number: 94055-76-2
MDL Number: MFCD00867604
Molecular Formula: C23H33NO7S2
Molecular Weight: 499.64
Purity/Analysis Method: >98.0% (HPLC,N)
Form: Crystal
Color: Very Pale Yellow
Melting point (°C): 87


Product Categories: Bioscience, Immune System, Inflammation, Allergy Research Reagents, Other Pharmaceutical Ingredients
Other Product Names / Synonyms: 2-[4-(3-Ethoxy-2-hydroxypropoxy)phenylcarbamoyl]ethyldimethylsulfonium p-Toluenesulfonate

Formula: C₂₃H₃₃NO₇S₂
Melting Pt: 87 °C
MDL Number: MFCD00867604
CAS Number: 94055-76-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bb1349aa22d8f2d27728ade212cca73b6a086c1e&vid=bb1349aa22d8f2d27728ade212cca73b6a086c1e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR