N-Succinimidyl 7-Methoxycoumarin-3-carboxylate ≥98.0% (by HPLC, total nitrogen)
Supplier: TCI America
Synonyms:
7-Methoxycoumarin-3-carboxylic Acid N-Succinimidyl Ester
CAS Number: 150321-92-9
MDL Number: MFCD00467593
Molecular Formula: C15H11NO7
Molecular Weight: 317.25
Purity/Analysis Method: >98.0% (HPLC,N)
Form: Crystal
Color: White
Storage Temperature: <0°C
Product Categories: Bioscience, Donors & Acceptors for Fluorescent Resonance Energy Transfer (FRET), Fluorescent Labeling Dyes, Analytical Science, Fluorescent Dyes
Other Product Names / Synonyms: 7-Methoxycoumarin-3-carboxylic Acid N-Succinimidyl Ester
Formula:
C₁₅H₁₁NO₇ MW: 317.25 g/mol |
MDL Number:
MFCD00467593 CAS Number: 150321-92-9 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...