N-Succinimidyl 7-Hydroxycoumarin-3-carboxylate ≥96.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: 2,5-Dioxopyrrolidin-1-yl 7-hydroxy-2-oxo-2H-chromene-3-carboxylate

S0866-1G S0866-200MG
TCS0866-1GEA 790.16 CAD
TCS0866-1G TCS0866-200MG
N-Succinimidyl 7-Hydroxycoumarin-3-carboxylate ≥96.0% (by HPLC, titration analysis)
N-Succinimidyl 7-Hydroxycoumarin-3-carboxylate

CAS Number: 134471-24-2
MDL Number: MFCD00037568
Molecular Formula: C14H9NO7
Molecular Weight: 303.23
Purity/Analysis Method: >96.0% (HPLC,T)
Form: Crystal
Color: Slightly Pale Yellow
Storage Temperature: <0°C


Product Categories: Bioscience, Donors & Acceptors for Fluorescent Resonance Energy Transfer (FRET), Fluorescent Labeling Dyes, Analytical Science, Fluorescent Dyes
Other Product Names / Synonyms: 7-Hydroxycoumarin-3-carboxylic Acid N-Succinimidyl Ester, N-Succinimidyl Umbelliferone-3-carboxylate, Umbelliferone-3-carboxylic Acid N-Succinimidyl Ester

Formula: C₁₄H₉NO₇
MW: 303.23 g/mol
MDL Number: MFCD00037568
CAS Number: 134471-24-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d903e618860485eea603bba15bf71a50275b52bf&vid=d903e618860485eea603bba15bf71a50275b52bf&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR