4-(Succinimidylcarboxy)benzophenone ≥98.0%

Supplier: TCI America

Synonyms: 4-Benzoylbenzoic acid N-succinimidyl ester, 2,5-Dioxopyrrolidin-1-yl 4-benzoylbenzoate

S0863-1G S0863-200MG
TCS0863-1GEA 1006.46 CAD
TCS0863-1G TCS0863-200MG
4-(Succinimidylcarboxy)benzophenone ≥98.0%
4-(Succinimidylcarboxy)benzophenone

CAS Number: 91990-88-4
MDL Number: MFCD00058572
Molecular Formula: C18H13NO5
Molecular Weight: 323.30
Purity/Analysis Method: >98.0% (HPLC,N)
Form: Crystal
Color: White
Melting point (°C): 209
Storage Temperature: 0-10°C


Product Categories: Bioscience, Crosslinkers
Other Product Names / Synonyms: 4-Benzoylbenzoic Acid N-Succinimidyl Ester

Formula: C₁₈H₁₃NO₄
Melting Pt: 209 °C
MDL Number: MFCD00058572
CAS Number: 91990-88-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9afd50e87d8c5ed0bb428c9ff42a5ab9a2bb2e28&vid=9afd50e87d8c5ed0bb428c9ff42a5ab9a2bb2e28&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR