Sodium-2-ethylhexanoate ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: 2-Ethylcaproic acid sodium salt, Sodium 2-ethylhexanoate, 2-Ethylhexanoic acid sodium salt

S0862-250G S0862-25G
TCS0862-250GEA 93.22 CAD
TCS0862-250G TCS0862-25G
Sodium-2-ethylhexanoate ≥98.0% (by titrimetric analysis)
Sodium 2-ethylhexanoate

CAS Number: 19766-89-3
MDL Number: MFCD00014007
Molecular Formula: C8H16O2
Molecular Weight: 166.20
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Color: White


Product Categories: Bioscience, Lipids
Other Product Names / Synonyms: 2-Ethylhexanoic Acid Sodium Salt, Octanoic Acid Sodium Salt, Sodium Octanoate

Formula: C₈H₁₅NaO₂
MW: 166.2 g/mol
MDL Number: MFCD00014007
CAS Number: 19766-89-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=54556574e9bd92c5702ce4a4eb17442c852d7f9e&vid=54556574e9bd92c5702ce4a4eb17442c852d7f9e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR