N-Succinimidyl 4-[4-(dimethylamino)phenylazo]benzoate ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
S0857-1G
S0857-200MG
TCS0857-1GEA
556.16
CAD
TCS0857-1G
TCS0857-200MG
N-Succinimidyl 4-[4-(dimethylamino)phenylazo]benzoate ≥98.0% (by HPLC, titration analysis)
N-Succinimidyl 4-[4-(dimethylamino)phenylazo]benzoate
CAS Number: 146998-31-4
MDL Number: MFCD00467415
Molecular Formula: C19H18N4O4
Molecular Weight: 366.38
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Color: Colorless
Melting point (°C): 242
Storage Temperature: 0-10°C
Product Categories: Bioscience, Donors & Acceptors for Fluorescent Resonance Energy Transfer (FRET)
Other Product Names / Synonyms: 4-[4-(Dimethylamino)phenylazo]benzoic Acid N-Succinimidyl Ester, DABCYL N-Succinimidyl Ester
Formula:
C₁₉H₁₈N₄O₄ Melting Pt: 242 °C |
MDL Number:
MFCD00467415 CAS Number: 146998-31-4 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...