Sorbic acid ≥99.0% (by titrimetric analysis) for biochemical research
Supplier: TCI America
Synonyms:
(E,E)-2,4-Hexadienoic acid, (2E,4E)-2,4-Hexadienoic acid, trans,trans-2,4-Hexadienoic acid, 2,4-Hexadienoic acid, (2E,4E)-Hexa-2,4-dienoic acid
[for Biochemical Research]
CAS Number: 110-44-1
MDL Number: MFCD00002703
Molecular Formula: C6H8O2
Molecular Weight: 112.13
Purity/Analysis Method: >99.0% (T)
Form: Crystal
Color: White
Boiling point (°C): 228
Melting point (°C): 134
Flash Point (°C): 127
Storage Temperature: 0-10°C
Product Categories: Bioscience, Preservatives for Biochemical Research
Other Product Names / Synonyms: 2,4-Hexadienoic Acid
Formula:
CH₃CH=CHCH=CHCOOH MW: 112.13 g/mol Boiling Pt: 228 Melting Pt: 134 °C Flash Pt: 127 °C |
MDL Number:
MFCD00002703 CAS Number: 110-44-1 Merck Index: 13,08793 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...