Salicylaldehyde azine ≥98.0%

Supplier: TCI America

Synonyms: Bis(2-hydroxybenzaldehyde) hydrazone, N,N'-Bis(salicylidene)hydrazine, Salazine, Salicylaldazine, 2,2'-(Hydrazine-1,2-diylidenebis(methanylylidene))diphenol

S0850-25G S0850-5G
TCS0850-25GEA 309.68 CAD
TCS0850-25G TCS0850-5G
Salicylaldehyde azine ≥98.0%
Salicylaldehyde azine

CAS Number: 959-36-4
MDL Number: MFCD00043496
Molecular Formula: C14H12N2O2
Molecular Weight: 240.26
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Color: Pale Yellow
Melting point (°C): 218


Product Categories: Materials Chemistry, Metal Organic Framework (MOF) and Porous Coordination Polymer (PCP) Research Reagents
Other Product Names / Synonyms: 2-Hydroxybenzaldehyde Azine, Salicylalazine, Salicylaldazine

Formula: C₁₄H₁₂N₂O₂
Melting Pt: 218 °C
MDL Number: MFCD00043496
CAS Number: 959-36-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ea4900a21732c40dd9256621d98b12aa092a73f6&vid=ea4900a21732c40dd9256621d98b12aa092a73f6&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR