Sialyl lewis-X-lactose ≥97.0% (by HPLC)

Supplier: TCI America

Synonyms: Neu5Ac alpha(2-3)Gal beta(1-4)[Fuc alpha(1-3)]GlcNAc beta(1-3)Gal beta(1-4)Glc

S0849-1MG S0849-5MG
TCS0849-1MGEA 811.65 CAD
TCS0849-1MG TCS0849-5MG
Sialyl lewis-X-lactose ≥97.0% (by HPLC)
Sialyl lewis-X-lactose

CAS Number: 127923-85-7
Molecular Formula: C43H72N2O33
Molecular Weight: 1145.03
Purity/Analysis Method: >97.0% (HPLC)
Form: Crystal
Color: White
Storage Temperature: <0°C


Product Categories: Bioscience, Sialylated Oligosaccharides, Oligosaccharides
Other Product Names / Synonyms: Neu5Ac alpha(2-3)Gal beta(1-4)[Fuc alpha(1-3)]GlcNAc beta(1-3)Gal beta(1-4)Glc

Formula: C₄₃H₇₂N₂O₃₃
MW: 1145.03 g/mol
CAS Number: 127923-85-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=daa433d61d9093518022d17c9fd00939453b681d&vid=daa433d61d9093518022d17c9fd00939453b681d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR