Sodium-2-iodobenzenesulfonate ≥97.0% (by HPLC)

Supplier: TCI America

S0848-1G S0848-5G
TCS0848-1GEA 175.22 CAD
TCS0848-1G TCS0848-5G
Sodium-2-iodobenzenesulfonate ≥97.0% (by HPLC)
Sodium-2-iodobenzenesulfonate

CAS Number: 62973-69-7
MDL Number: MFCD12031329
Molecular Formula: C6H5IO3S
Molecular Weight: 306.05
Purity/Analysis Method: >97.0% (HPLC)
Form: Crystal
Color: White


Product Categories: Synthetic Organic Chemistry, Sulfonic Acid Derivatives
Other Product Names / Synonyms: 2-Iodobenzenesulfonic Acid Sodium Salt

Formula: C₆H₅IO₃S
MDL Number: MFCD12031329
CAS Number: 62973-69-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8230788aa0e34c98e4da715d0b2a9dee5faba0a0&vid=8230788aa0e34c98e4da715d0b2a9dee5faba0a0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR