Sarcosine ethyl ester hydrochloride ≥98.0%
Supplier: TCI America
Synonyms:
Sarcosine Ethyl Ester HCl, H-Sar-OEt·HCl, Ethyl 2-(methylamino)acetate hydrochloride
CAS Number: 52605-49-9
MDL Number: MFCD00012506
Molecular Formula: C5H11NO2
Molecular Weight: 153.61
Purity/Analysis Method: >98.0% (N,T)
Form: Crystal
Color: White
Melting point (°C): 125
Product Categories: Bioscience, Biologically-modified Amino Acids, Amino Acids (C-Protected)
Other Product Names / Synonyms: Ethyl (Methylamino)acetate Hydrochloride, N-Methylglycine Ethyl Ester Hydrochloride, Me-Gly-OEt.HCl
Formula:
C₅H₁₂ClNO₂ Melting Pt: 125 °C |
MDL Number:
MFCD00012506 CAS Number: 52605-49-9 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...