Sucralose ≥98.0%

Supplier: TCI America

Synonyms: Trichlorosucrose, (2R,3R,4R,5R,6R)-2-(((2R,3S,4S,5S)-2,5-Bis(chloromethyl)-3,4-dihydroxytetrahydrofuran-2-yl)oxy)-5-chloro-6-(hydroxymethyl)tetrahydro-2H-pyran-3,4-diol, 1-(1,6-Dichloro-1,6-dideoxy-beta-D-fructofuranosyl)-4-chloro-4-deoxy-alpha-D-galactopyranoside

S0839-25G S0839-5G S0839-100G
TCS0839-25GEA 193.2 CAD
TCS0839-25G TCS0839-5G TCS0839-100G
Sucralose ≥98.0%
Sucralose

CAS Number: 56038-13-2 MDL Number: MFCD03648615 Molecular Formula: C12H19Cl3O8 Molecular Weight: 397.63 Purity/Analysis Method: 98.0% (HPLC) Form: Crystal Color: White Melting point (°C): 130 Specific rotation [a]20/D: 86.0 deg (C=1, H2O)


Form: Crystal
Color: White
Product Categories: Bioscience, Obesity/Diabete Research Reagents, Disaccharides
Other Product Names / Synonyms: 1-(1,6-Dichloro-1,6-dideoxy-beta-D-fructofuranosyl)-4-chloro-4-deoxy-alpha-D-galactopyranoside

Formula: C₁₂H₁₉Cl₃O₈
MW: 397.63 g/mol
Melting Pt: 130 °C
MDL Number: MFCD03648615
CAS Number: 56038-13-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e43c69636674865ce3859fa4b8ae5663bfcaad77&vid=e43c69636674865ce3859fa4b8ae5663bfcaad77&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR