Sodium dicyanoamide ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: Sodium dicyanamide

S0838-25G S0838-100G
TCS0838-25GEA 67.53 CAD
TCS0838-25G TCS0838-100G
Sodium dicyanoamide ≥98.0% (by titrimetric analysis)
Sodium dicyanoamide

CAS Number: 1934-75-4
MDL Number: MFCD00003535
Molecular Formula: C2N3Na
Molecular Weight: 89.03
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Color: White
Melting point (°C): 260

Formula: C₂N₃Na
MW: 89.03 g/mol
Melting Pt: 260 °C
MDL Number: MFCD00003535
CAS Number: 1934-75-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2991366ac3d840852a18864e44d6fd46e2279272&vid=2991366ac3d840852a18864e44d6fd46e2279272&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR