Sodium-2,3,5,6-tetrafluoro-4-hydroxybenzenesulfonate ≥98.0% (by HPLC)

Supplier: TCI America

S0836-1G S0836-5G
TCS0836-1GEA 272.71 CAD
TCS0836-1G TCS0836-5G
Sodium-2,3,5,6-tetrafluoro-4-hydroxybenzenesulfonate ≥98.0% (by HPLC)
Sodium-2,3,5,6-tetrafluoro-4-hydroxybenzenesulfonate

CAS Number: 221908-17-4
Molecular Formula: C6H2F4O4S
Molecular Weight: 268.11
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Color: White


Product Categories: Synthetic Organic Chemistry, Condensation & Active Esterification, Fluorinated Building Blocks
Other Product Names / Synonyms: 2,3,5,6-Tetrafluoro-4-hydroxybenzenesulfonic Acid Sodium Salt

Formula: C₆H₂F₄O₄S
CAS Number: 221908-17-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8c144789df08295b3024045f32ac2a8b9ea21b57&vid=8c144789df08295b3024045f32ac2a8b9ea21b57&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR