9,9'-Spirobi[9H-fluorene]-2-boronic acid ≥95.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: 9,9'-Spirobifluorene-2-boronic acid

S0831-1G S0831-5G
TCS0831-1GEA 74.26 CAD
TCS0831-1G TCS0831-5G
9,9'-Spirobi[9H-fluorene]-2-boronic acid ≥95.0% (by titrimetric analysis)
9,9'-Spirobi[9H-fluorene]-2-boronic acid

CAS Number: 236389-21-2
Molecular Formula: C25H17BO2
Molecular Weight: 360.22
Form: Crystal
Color: White

Formula: C₂₅H₁₇BO₂
MW: 360.22 g/mol
MDL Number: MFCD13248568
CAS Number: 236389-21-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=53ed4c846127a67ba3a353baba830890dd292dce&vid=53ed4c846127a67ba3a353baba830890dd292dce&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR