Sodium-3-(N-ethyl-3-methoxyanilino)-2-hydroxy-1-propanesulfonate ≥98.0% (by HPLC, titration analysis) for biochemical research
Supplier: TCI America
Synonyms:
3-(N-Ethyl-3-methoxyanilino)-2-hydroxy-1-propanesulfonic acid Sodium Salt, ADOS, N-Ethyl-N-(2-hydroxy-3-sulfopropyl)-3-methoxyaniline Sodium Salt, 3-(N-Ethyl-3-methoxyanilino)-2-hydroxy-1-propanesulphonic acid Sodium Salt, N-Ethyl-N-(2-hydroxy-3-sulphopropyl)-3-methoxyaniline Sodium Salt
[for Biochemical Research]
CAS Number: 82692-96-4
MDL Number: MFCD09261431
Molecular Formula: C12H19NO5S
Molecular Weight: 311.33
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Color: White
Melting point (°C): 144
Product Categories: Bioscience, Peroxidase Substrates, Trinder's Reagents
Other Product Names / Synonyms: 3-(N-Ethyl-3-methoxyanilino)-2-hydroxy-1-propanesulfonic Acid Sodium Salt, N-Ethyl-N-(2-hydroxy-3-sulfopropyl)-3-methoxyaniline Sodium Salt, ADOS
Formula:
C₁₂H₁₉NO₅S Melting Pt: 144 °C |
MDL Number:
MFCD09261431 CAS Number: 82692-96-4 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...