Sodium-3-(N-ethylanilino)propanesulfonate ≥98.0% (by HPLC) for biochemical research
Supplier: TCI America
Synonyms:
N-Ethyl-N-(3-sulphopropyl)aniline Sodium Salt, ALPS, 3-(N-Ethylanilino)propanesulphonic acid Sodium Salt, Sodium 3-(ethyl(phenyl)amino)propane-1-sulphonate, 3-(N-Ethylanilino)propanesulfonic acid Sodium Salt, N-Ethyl-N-(3-sulfopropyl)aniline Sodium Salt, Sodium 3-(ethyl(phenyl)amino)propane-1-sulfonate
[for Biochemical Research]
CAS Number: 82611-85-6
MDL Number: MFCD01311047
Molecular Formula: C11H17NO3S
Molecular Weight: 265.30
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Color: White
Product Categories: Bioscience, Peroxidase Substrates, Trinder's Reagents
Other Product Names / Synonyms: 3-(N-Ethylanilino)propanesulfonic Acid Sodium Salt, N-Ethyl-N-(3-sulfopropyl)aniline Sodium Salt, ALPS
Formula:
C₁₁H₁₇NO₃S MW: 265.30 g/mol |
MDL Number:
MFCD01311047 CAS Number: 82611-85-6 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...