1-(3-sulfopropyl)pyridinium hydroxide inner salt ≥98.0% (by HPLC, total nitrogen) for biochemical research
Supplier: TCI America
Synonyms:
NDSB 201, 3-(1-Pyridinio)propanesulfonate, 3-(1-Pyridinio)propanesulphonate, 3-(Pyridin-1-ium-1-yl)propane-1-sulphonate, 3-(Pyridin-1-ium-1-yl)propane-1-sulfonate
[for Biochemical Research]
CAS Number: 15471-17-7
MDL Number: MFCD00064468
Molecular Formula: C8H11NO3S
Molecular Weight: 201.24
Purity/Analysis Method: >98.0% (HPLC,N)
Form: Crystal
Color: White
Melting point (°C): 275
Flash Point (°C): 160
Product Categories: Bioscience, Non-detergent Sulfobetaines (NDSB)
Other Product Names / Synonyms: 3-(1-Pyridinio)propanesulfonate, NDSB 201
Formula:
C₈H₁₁NO₃S MW: 201.24 g/mol Melting Pt: 275 °C Flash Pt: 160 °C |
MDL Number:
MFCD00064468 CAS Number: 15471-17-7 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...