Sulfacetamide ≥98.0%

Supplier: TCI America

Synonyms: N-((4-Aminophenyl)sulfonyl)acetamide, N-((4-Aminophenyl)sulphonyl)acetamide

S0577-250G S0577-25G
TCS0577-250GEA 257.54 CAD
TCS0577-250G TCS0577-25G
Sulfacetamide ≥98.0%
Sulphacetamide

CAS Number: 144-80-9
Molecular Formula: C8H10N2O3S
Molecular Weight: 214.24
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Color: White
Melting point (°C): 183


Product Categories: Bioscience, Synthetic Antimicrobial Ingredients
Other Product Names / Synonyms: N-Acetyl-4-aminobenzenesulfonamide, N-Acetylsulfanilamide

Formula: C₈H₁₀N₂O₃S
Melting Pt: 183 °C
MDL Number: MFCD00066501
CAS Number: 144-80-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d65169aa764ef06070e91d926d569269e18d32ed&vid=d65169aa764ef06070e91d926d569269e18d32ed&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR