N-Succinimidyl 6-Biotinamidohexanoate ≥97.0% (by HPLC)
Supplier: TCI America
S0490-100MG
S0490-20MG
TCS0490-100MGEA
251.22
CAD
TCS0490-100MG
TCS0490-20MG
N-Succinimidyl 6-Biotinamidohexanoate ≥97.0% (by HPLC)
N-Succinimidyl 6-Biotinamidohexanoate
CAS Number: 72040-63-2
MDL Number: MFCD00065502
Molecular Formula: C20H30N4O6S
Molecular Weight: 454.54
Purity/Analysis Method: >97.0% (HPLC)
Form: Crystal
Color: White
Melting point (°C): 170
Storage Temperature: <0°C
Product Categories: Bioscience, Biotins & Streptavidins
Other Product Names / Synonyms: 6-Biotinamidohexanoic Acid N-Succinimidyl Ester, N-[5-(N-Succinimidyloxycarbonyl)pentyl]biotinamide, N-Biotinyl-6-aminohexanoic Acid Succinimidyl Ester
Formula:
C₂₀H₃₀N₄O₆S Melting Pt: 170 °C |
MDL Number:
MFCD00065502 CAS Number: 72040-63-2 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...