Sodium borohydride ≥95.0% (by titrimetric analysis)

Supplier: TCI America

S0480-100G S0480-500G S0480-25G
TCS0480-100GEA 135.94 CAD
TCS0480-100G TCS0480-500G TCS0480-25G
Sodium borohydride ≥95.0% (by titrimetric analysis)
Sodium borohydride

CAS Number: 16940-66-2
MDL Number: MFCD00003518
Molecular Formula: H4BNa
Molecular Weight: 37.83
Purity/Analysis Method: >95.0% (T)
Form: Crystal
Color: White


Product Categories: Synthetic Organic Chemistry, Reduction, Materials Chemistry, Fuel Cell Research Reagents
Other Product Names / Synonyms: Sodium Tetrahydroborate, Sodium Tetrahydridoborate

Formula: NaBH₄
MDL Number: MFCD00003518
CAS Number: 16940-66-2
UN: 1426
ADR: 4.3,I
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=53f98a43cd5cd3b0e549ff43068403b1a1ce4ae0&vid=53f98a43cd5cd3b0e549ff43068403b1a1ce4ae0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR