Sodium tetrakis[3,5-bis(trifluoromethyl)phenyl]borate hydrate ≥98.0% (by HPLC and gravimetric analysis)
Supplier: TCI America
Synonyms:
NaBARF
S0447-1G
S0447-200MG
TCS0447-1GEA
346.7
CAD
TCS0447-1G
TCS0447-200MG
Sodium tetrakis[3,5-bis(trifluoromethyl)phenyl]borate hydrate ≥98.0% (by HPLC and gravimetric analysis)
Sodium tetrakis[3,5-bis(trifluoromethyl)phenyl]borate hydrate
CAS Number: 79060-88-1
MDL Number: MFCD00043323
Molecular Formula: C32H12BF24Na
Molecular Weight: 886.21
Purity/Analysis Method: >98.0% (HPLC,W)
Form: Crystal
Color: Very Pale Yellow
Other Product Names / Synonyms: NaBARF
Formula:
C₃₂H₁₂BF₂₄Na.xH₂O MW: 886.21 g/mol (anhy) |
MDL Number:
MFCD00043323 CAS Number: 79060-88-1 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more
![](/m-ca.vwr.com/en_CA/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...