Diethanolamine 99,0

Supplier: TCI America

Synonyms: ß,ß′-Dihydroxydiethylamine, Bis-(ß-hydroxyethyl)-amine, 2,2'-Iminodiethanol

S0376-10G S0376-1G
TCS0376-10GEA 163.69 CAD
TCS0376-10G TCS0376-1G
Diethanolamine 99,0
Diethanolamine

[Matrix for FABMS and liquid SIMS]
CAS Number: 111-42-2
MDL Number: MFCD00002843
Molecular Formula: C4H11NO2
Molecular Weight: 105.14
Purity/Analysis Method: >99.0% (GC)
Form: Crystal
Color: White
Flash Point (°C): 160
Freezing point (°C): 27


Product Categories: Analytical Science, Standards for Mass Spectrometry & Matrix Materials
Other Product Names / Synonyms: 2,2'-Dihydroxydiethylamine, 2,2'-Iminodiethanol

Formula: C₄H₁₁NO₂
MW: 105.14 g/mol
Boiling Pt: 268.8
Melting Pt: 28
Flash Pt: 160 °C
MDL Number: MFCD00002843
CAS Number: 111-42-2
Merck Index: 13,03134
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0ce3f8143e4cdb7982ec30d1fcad1f5ab170fe0c&vid=b3f470785a1beb1ddff0dc0ad9841c3521183beb&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR