[RuCl(p-cymene)((S)-dm-segphos®)]Cl

Supplier: TCI America

Synonyms: Chloro{(S)-(-)-5,5'-bis[di(3,5-xylyl)phosphino]-4,4'-bi-1,3-benzodioxole}(p-cymene)ruthenium(II) chloride [RuCl(p-cymene)((S)-dm-segphos®)]Cl, Chloro[(S)-(-)-5,5'-bis[di(3,5-xylyl)phosphino]-4,4'-bi-1,3-benzodioxole](p-cymene)ruthenium(II) Chloride

R0157-1G R0157-200MG
TCR0157-1GEA 317.42 CAD
TCR0157-1G TCR0157-200MG
[RuCl(p-cymene)((S)-dm-segphos®)]Cl
[RuCl(p-cymene)((S)-dm-segphos®)]Cl

CAS Number: 944451-31-4
MDL Number: MFCD09753018
Molecular Formula: C56H58Cl2O4P2Ru
Molecular Weight: 1028.99
Form: Crystal
Color: Yellow


Other Product Names / Synonyms: Chloro[(S)-(-)-5,5'-bis[di(3,5-xylyl)phosphino]-4,4'-bi-1,3-benzodioxole](p-cymene)ruthenium(II) Chloride

Formula: C₅₆H₅₈Cl₂O₄P₂Ru
MW: 1028.99 g/mol
MDL Number: MFCD09753018
CAS Number: 944451-31-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1ff5dcb273afc37b4416e9d4f9b2e4edf1a9fe9d&vid=1ff5dcb273afc37b4416e9d4f9b2e4edf1a9fe9d&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR