[RuCl(p-cymene)((R)-dm-segphos®)]Cl
Supplier: TCI America
R0156-1G
R0156-200MG
TCR0156-1GEA
317.42
CAD
TCR0156-1G
TCR0156-200MG
[RuCl(p-cymene)((R)-dm-segphos®)]Cl
[RuCl(p-cymene)((R)-dm-segphos®)]Cl
CAS Number: 944451-30-3
MDL Number: MFCD09753018
Molecular Formula: C56H58Cl2O4P2Ru
Molecular Weight: 1028.99
Form: Crystal
Color: Yellow
Other Product Names / Synonyms: Chloro[(R)-(+)-5,5'-bis[di(3,5-xylyl)phosphino]-4,4'-bi-1,3-benzodioxole](p-cymene)ruthenium(II) Chloride
Formula:
C₅₆H₅₈Cl₂O₄P₂Ru |
MDL Number:
MFCD09753018 CAS Number: 944451-30-3 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...