[RuCl(p-cymene)((S)-segphos®)]Cl

Supplier: TCI America

R0155-1G R0155-200MG
TCR0155-1GEA 274.45 CAD
TCR0155-1G TCR0155-200MG
[RuCl(p-cymene)((S)-segphos®)]Cl
[RuCl(p-cymene)((S)-segphos®)]Cl

CAS Number: 944451-29-0
MDL Number: MFCD09753017
Molecular Formula: C48H42Cl2O4P2Ru
Molecular Weight: 916.78
Form: Crystal
Color: Reddish Yellow


Other Product Names / Synonyms: Chloro[(S)-(-)-5,5'-bis(diphenylphosphino)-4,4'-bi-1,3-benzodioxole](p-cymene)ruthenium(II) Chloride

Formula: C₄₈H₄₂Cl₂O₄P₂Ru
MDL Number: MFCD09753017
CAS Number: 944451-29-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f98932c8df12177777dd342acad7327a0532b56f&vid=f98932c8df12177777dd342acad7327a0532b56f&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR