[RuCl(p-cymene)((R)-segphos®)]Cl

Supplier: TCI America

Synonyms: Chloro[(R)-(+)-5,5'-bis(diphenylphosphino)-4,4'-bi-1,3-benzodioxole](p-cymene)ruthenium(II) chloride [RuCl(p-cymene)((R)-segphos®)]Cl, Chloro[(R)-(+)-5,5'-bis(diphenylphosphino)-4,4'-bi-1,3-benzodioxole](p-cymene)ruthenium(II) Chloride

R0154-1G R0154-200MG
TCR0154-1GEA 216.46 CAD
TCR0154-1G TCR0154-200MG
[RuCl(p-cymene)((R)-segphos®)]Cl
[RuCl(p-cymene)((R)-segphos®)]Cl

CAS Number: 944451-28-9
MDL Number: MFCD09753017
Molecular Formula: C48H42Cl2O4P2Ru
Molecular Weight: 916.78
Form: Crystal
Color: Reddish Yellow


Other Product Names / Synonyms: Chloro[(R)-(+)-5,5'-bis(diphenylphosphino)-4,4'-bi-1,3-benzodioxole](p-cymene)ruthenium(II) Chloride

Formula: C₄₈H₄₂Cl₂O₄P₂Ru
MW: 916.78 g/mol
MDL Number: MFCD09753017
CAS Number: 944451-28-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6203c8c0e47bac7f934236dbd38da5140400c68d&vid=6203c8c0e47bac7f934236dbd38da5140400c68d&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR