[RuCl(p-cymene)((S)-xylbinap)]Cl

Supplier: TCI America

R0151-1G R0151-200MG
TCR0151-1GEA 322.32 CAD
TCR0151-1G TCR0151-200MG
[RuCl(p-cymene)((S)-xylbinap)]Cl
[RuCl(p-cymene)((S)-xylbinap)]Cl

CAS Number: 944451-25-6
MDL Number: MFCD09753015
Molecular Formula: C62H62Cl2P2Ru
Molecular Weight: 1041.10
Form: Crystal
Color: Yellow


Other Product Names / Synonyms: Chloro[(S)-(-)-2,2'-bis[di(3,5-xylyl)phosphino]-1,1'-binaphthyl](p-cymene)ruthenium(II) Chloride

Formula: C₆₂H₆₂Cl₂P₂Ru
MDL Number: MFCD09753015
CAS Number: 944451-25-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=306840f14c34f0f3701d7924f6b21afa3e3f71fd&vid=306840f14c34f0f3701d7924f6b21afa3e3f71fd&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR