[RuCl(p-cymene)((S)-tolbinap)]Cl

Supplier: TCI America

R0149-1G R0149-200MG
TCR0149-1GEA 241.27 CAD
TCR0149-1G TCR0149-200MG
[RuCl(p-cymene)((S)-tolbinap)]Cl
[RuCl(p-cymene)((S)-tolbinap)]Cl

CAS Number: 228120-95-4
MDL Number: MFCD09753014
Molecular Formula: C58H54Cl2P2Ru
Molecular Weight: 984.99
Form: Crystal
Color: Yellow


Other Product Names / Synonyms: Chloro[(S)-(-)-2,2'-bis(di-p-tolylphosphino)-1,1'-binaphthyl](p-cymene)ruthenium(II) Chloride

Formula: C₅₈H₅₄Cl₂P₂Ru
MDL Number: MFCD09753014
CAS Number: 228120-95-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5a75b5d915ce1b365cf25641660c866d796a8c68&vid=5a75b5d915ce1b365cf25641660c866d796a8c68&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR