[RuCl(p-cymene)((S)-binap)]Cl ≥90.0% (by HPLC)

Supplier: TCI America

R0147-1G R0147-200MG
TCR0147-1GEA 249.64 CAD
TCR0147-1G TCR0147-200MG
[RuCl(p-cymene)((S)-binap)]Cl ≥90.0% (by HPLC)
[RuCl(p-cymene)((S)-binap)]Cl

CAS Number: 130004-33-0
MDL Number: MFCD00134456
Molecular Formula: C54H46Cl2P2Ru
Molecular Weight: 928.88
Purity/Analysis Method: >90.0% (HPLC)
Form: Crystal
Color: Pale Reddish Yellow


Product Categories: Synthetic Organic Chemistry, Homogeneous Catalysts, Asymmetric Synthesis
Other Product Names / Synonyms: Chloro[(S)-(-)-2,2'-bis(diphenylphosphino)-1,1'-binaphthyl](p-cymene)ruthenium(II) Chloride

Formula: C₅₄H₄₆Cl₂P₂Ru
MDL Number: MFCD00134456
CAS Number: 130004-33-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5b4c1c382683887767bb9fc7644a36b08e7e23d8&vid=5b4c1c382683887767bb9fc7644a36b08e7e23d8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 90.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR