[RuCl(p-cymene)((R)-binap)]Cl ≥90.0%

Supplier: TCI America

Synonyms: Chloro[(R)-(+)-2,2'-bis(diphenylphosphino)-1,1'-binaphthyl](p-cymene)ruthenium(II) chloride [RuCl(p-cymene)((R)-binap)]Cl, (R)-RuCl[(p-cymene)(BINAP)]Cl, Chloro[(R)-(+)-2,2'-bis(diphenylphosphino)-1,1'-binaphthyl](p-cymene)ruthenium(II) Chloride, [(R)-Binap RuCl p-cymene]Cl

R0146-1G R0146-200MG
TCR0146-1GEA 218.2 CAD
TCR0146-1G TCR0146-200MG
[RuCl(p-cymene)((R)-binap)]Cl ≥90.0%
[RuCl(p-cymene)((R)-binap)]Cl

CAS Number: 145926-28-9
MDL Number: MFCD00134456
Molecular Formula: C54H46Cl2P2Ru
Molecular Weight: 928.88
Purity/Analysis Method: >90.0% (HPLC)
Form: Crystal
Color: Pale Reddish Yellow


Product Categories: Synthetic Organic Chemistry, Homogeneous Catalysts, Asymmetric Synthesis
Other Product Names / Synonyms: Chloro[(R)-(+)-2,2'-bis(diphenylphosphino)-1,1'-binaphthyl](p-cymene)ruthenium(II) Chloride

Formula: C₅₄H₄₆Cl₂P₂Ru
MW: 928.88 g/mol
MDL Number: MFCD00134456
CAS Number: 145926-28-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6d9c9b3d85eb589b136423a752a33d12237c73e0&vid=6d9c9b3d85eb589b136423a752a33d12237c73e0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 90.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR