Ru-MACHO®-BH

Supplier: TCI America

Synonyms: Carbonylhydrido(tetrahydroborato)[bis(2-diphenylphosphinoethyl)amino]ruthenium(II)

R0137-1G R0137-200MG
TCR0137-1GEA 358.69 CAD
TCR0137-1G TCR0137-200MG
Ru-MACHO®-BH
Ru-MACHO®-BH

CAS Number: 1295649-41-0
MDL Number: MFCD20922901
Molecular Formula: C29H34BNOP2Ru
Molecular Weight: 586.42
Form: Crystal
Color: White


Other Product Names / Synonyms: Carbonylhydrido(tetrahydroborato)[bis(2-diphenylphosphinoethyl)amino]ruthenium(II)

Formula: C₂₉H₃₄BNOP₂Ru
MW: 586.42 g/mol
MDL Number: MFCD20922901
CAS Number: 1295649-41-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5c6e33ae1ca2aa9781cfd080df133858ad29e625&vid=5c6e33ae1ca2aa9781cfd080df133858ad29e625&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR