Ru-MACHO® (contains 5% Toluene at maximum)

Supplier: TCI America

Synonyms: Carbonylchlorohydrido[bis(2-diphenylphosphinoethyl)amino]ruthenium(II)

R0136-1G R0136-200MG
TCR0136-1GEA 322.32 CAD
TCR0136-1G TCR0136-200MG
Ru-MACHO® (contains 5% Toluene at maximum)
Ru-MACHO®

CAS Number: 1295649-40-9
MDL Number: MFCD22377801
Molecular Formula: C29H30ClNOP2Ru
Molecular Weight: 607.03
Form: Crystal
Color: White


Other Product Names / Synonyms: Carbonylchlorohydrido[bis(2-diphenylphosphinoethyl)amino]ruthenium(II)

Formula: C₂₉H₃₀ClNOP₂Ru
MW: 607.03 g/mol
MDL Number: MFCD22377801
CAS Number: 1295649-40-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a9d7bfa2003ed67acc23c38f678a09fb54e97da5&vid=a9d7bfa2003ed67acc23c38f678a09fb54e97da5&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR