RuCl2[(R)-xylbinap][(R,R)-dpen]
Supplier: TCI America
Synonyms:
Dichloro[(R)-2,2'-bis[di(3,5-xylyl)phosphino]-1,1'-binaphthyl][(R,R)-1,2-diphenylethylenediamine]ruthenium(II), Dichloro{(R)-(+)-2,2'-bis[di(3,5-xylyl)phosphino]-1,1'-binaphthyl}[(1R,2R)-(+)-1,2-diphenylethylenediamine]ruthenium(II) RuCl2[(R)-xylbinap][(R,R)-dpen]
CAS Number: 220114-38-5
MDL Number: MFCD09753028
Molecular Formula: C66H64Cl2N2P2Ru
Molecular Weight: 1119.17
Form: Crystal
Color: Yellow
Other Product Names / Synonyms: Dichloro[(R)-2,2'-bis[di(3,5-xylyl)phosphino]-1,1'-binaphthyl][(R,R)-1,2-diphenylethylenediamine]ruthenium(II)
Formula:
C₆₆H₆₄Cl₂N₂P₂Ru MW: 1119.17 g/mol |
MDL Number:
MFCD09753028 CAS Number: 220114-38-5 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...