RuCl2[(S)-xylbinap][(S,S)-dpen]
Supplier: TCI America
Synonyms:
Dichloro{(S)-(-)-2,2'-bis[di(3,5-xylyl)phosphino]-1,1'-binaphthyl}[(1S,2S)-(-)-1,2-diphenylethylenediamine]ruthenium(II) RuCl2[(S)-xylbinap][(S,S)-dpen], Dichloro[(S)-2,2'-bis[di(3,5-xylyl)phosphino]-1,1'-binaphthyl][(S,S)-1,2-diphenylethylenediamine]ruthenium(II)
CAS Number: 220114-03-4
MDL Number: MFCD09753030
Molecular Formula: C66H64Cl2N2P2Ru
Molecular Weight: 1119.17
Form: Crystal
Color: Yellow
Other Product Names / Synonyms: Dichloro[(S)-2,2'-bis[di(3,5-xylyl)phosphino]-1,1'-binaphthyl][(S,S)-1,2-diphenylethylenediamine]ruthenium(II)
Formula:
C₆₆H₆₄Cl₂N₂P₂Ru MW: 1119.17 g/mol |
MDL Number:
MFCD09753030 CAS Number: 220114-03-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...