RuCl2[(R)-xylbinap][(R)-daipen]

Supplier: TCI America

R0133-1G R0133-200MG
TCR0133-1GEA 404.48 CAD
TCR0133-1G TCR0133-200MG
RuCl2[(R)-xylbinap][(R)-daipen]
RuCl2[(R)-xylbinap][(R)-daipen]

CAS Number: 220114-32-9
MDL Number: MFCD09753026
Molecular Formula: C71H74Cl2N2O2P2Ru
Molecular Weight: 1221.30
Form: Crystal
Color: Yellow

Formula: C₇₁H₇₄Cl₂N₂O₂P₂Ru
MDL Number: MFCD09753026
CAS Number: 220114-32-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=31f795baa764f56a74c80169ea83b87790fdfbdd&vid=31f795baa764f56a74c80169ea83b87790fdfbdd&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR