RuCl2[(S)-xylbinap][(S)-daipen]
Supplier: TCI America
Synonyms:
Dichloro{(S)-(-)-2,2'-bis[di(3,5-xylyl)phosphino]-1,1'-binaphthyl}[(2S)-(+)-1,1-bis(4-methoxyphenyl)-3-methyl-1,2-butanediamine]ruthenium(II) RuCl2[(S)-xylbinap][(S)-daipen]
R0132-1G
R0132-200MG
TCR0132-1GEA
403.37
CAD
TCR0132-1G
TCR0132-200MG
RuCl2[(S)-xylbinap][(S)-daipen]
RuCl2[(S)-xylbinap][(S)-daipen]
CAS Number: 220114-01-2
MDL Number: MFCD09753027
Molecular Formula: C71H74Cl2N2O2P2Ru
Molecular Weight: 1221.30
Form: Crystal
Color: Yellow
Formula:
C₇₁H₇₄Cl₂N₂O₂P₂Ru MW: 1221.30 g/mol |
MDL Number:
MFCD09753027 CAS Number: 220114-01-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...