RuCl2[(R)-dm-segphos®][(R,R)-dpen]

Supplier: TCI America

Synonyms: Dichloro{(R)-(+)-5,5'-bis[di(3,5-xylyl)phosphino]-4,4'-bi-1,3-benzodioxole}[(1R,2R)-(+)-1,2-diphenylethylenediamine]ruthenium(II) RuCl2[(R)-dm-segphos®][(R,R)-dpen]

R0131-1G R0131-200MG
TCR0131-1GEA 320.74 CAD
TCR0131-1G TCR0131-200MG
RuCl2[(R)-dm-segphos®][(R,R)-dpen]
RuCl2[(R)-dm-segphos®][(R,R)-dpen]

CAS Number: 944450-45-7
MDL Number: MFCD09753031
Molecular Formula: C60H60Cl2N2O4P2Ru
Molecular Weight: 1107.07
Form: Crystal
Color: Yellow

Formula: C₆₀H₆₀Cl₂N₂O₄P₂Ru
MW: 1107.07 g/mol
MDL Number: MFCD09753031
CAS Number: 944450-45-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8e5d6bfc1966bf56947610da0498a7c686e2cebc&vid=8e5d6bfc1966bf56947610da0498a7c686e2cebc&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR